4-Amino-2,3-xylenol ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAA0961-10GEA
2340
DKK
TCIAA0961-10G
4-Amino-2,3-xylenol ≥98.0% (by GC, titration analysis)
4-Amino-2,3-xylenol
Formel:
C₈H₁₁NO |
MDL Number:
MFCD00070554 CAS-nummer: 3096-69-3 EINECS: 221-446-4 |
Specifikationer Testresultater
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point (Decomposition) | 170.0 to 177.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...