Dodecyl chloroformate ≥90.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAF0223-25GEA
517
DKK
TCIAF0223-25G
Dodecyl chloroformate ≥90.0% (by GC)
Dodecyl chloroformate
Formel:
C₁₃H₂₅ClO₂ Molekylvægt: 248,7894 g/mol Kogepunkt: 168°C |
MDL Number:
MFCD00059483 CAS-nummer: 24460-74-0 EINECS: 246-272-6 UN nummer: 3277 ADR: 6.1,II |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...