Ethyl chlorofluoroacetate ≥98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A14196.14EA 1990 DKK
A14196.14
Ethyl chlorofluoroacetate ≥98%
Ethyl chlorofluoroacetate
Formel: C₄H₆ClFO₂
Molekylvægt: 140,54 g/mol
Kogepunkt: 128…130 °C
Massefylde: 1,225
Flammepunkt: 44 °C (111 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00039333
CAS-nummer: 401-56-9
EINECS: 206-930-5
UN nummer: 2920
ADR: 8,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR