Diheptylamine ≥98.0% (by GC, titration analysis)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

TCIAD2675-5GEA 2180 DKK
TCIAD2675-5G
Diheptylamine ≥98.0% (by GC, titration analysis)
Diheptylamine
Formel: C₁₄H₃₁N
Molekylvægt: 213,4026 g/mol
Kogepunkt: 148°C
MDL Number: MFCD00027309
CAS-nummer: 2470-68-0
UN nummer: 2735
ADR: 8,III
https://dk.vwr.com/stibo/web/std.lang.all/96/88/28979688.jpg

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR