4-Amino-2,3,5,6-tetrafluoropyridine ≥97.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

TCIAA1546-1GEA 304 DKK
TCIAA1546-1G
4-Amino-2,3,5,6-tetrafluoropyridine ≥97.0% (by GC)
4-Amino-2,3,5,6-tetrafluoropyridine
Formel: C₅H₂F₄N₂
Molekylvægt: 166,08
MDL Number: MFCD00010855
CAS-nummer: 1682-20-8
https://dk.vwr.com/stibo/web/std.lang.all/96/42/28929642.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 85.0 to 88.0 deg-C
Solubility in N,N-DMF very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR