Benzyl-4-fluorophenyl ketone ≥98.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAB3213-25GEA 821 DKK
TCIAB3213-25G
Benzyl-4-fluorophenyl ketone ≥98.0% (by GC)
Benzyl-4-fluorophenyl ketone
Formel: C₁₄H₁₁FO
MDL Number: MFCD00017960
CAS-nummer: 347-84-2
https://dk.vwr.com/stibo/web/std.lang.all/46/36/28934636.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR