4'-Ethylpropiophenone ≥96.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAE0461-25MLEA
394
DKK
TCIAE0461-25ML
TCIAE0461-500ML
4'-Ethylpropiophenone ≥96.0% (by GC)
4'-Ethylpropiophenone
Formel:
C₁₁H₁₄O Kogepunkt: 246°C |
MDL Number:
MFCD00191646 CAS-nummer: 27465-51-6 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...