Dimethylthexylchlorosilane ≥95.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAT2116-5GEA
714
DKK
TCIAT2116-5G
TCIAT2116-25G
Dimethylthexylchlorosilane ≥95.0% (by GC)
Dimethylthexylchlorosilane
Formel:
C₈H₁₉ClSi Kogepunkt: 56°C Flammepunkt: 46°C |
MDL Number:
MFCD00012175 CAS-nummer: 67373-56-2 UN nummer: 2986 ADR: 8,II |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...