Disulphoton sulphoxide
Leverandør: EHRENSTORFER GMBH
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERCA12985500EA
1520
DKK
EHERCA12985500
Disulphoton sulphoxide
Disulphoton sulphoxide
Formel:
C₈H₁₉O₃PS₃ |
CAS-nummer:
2497-07-6 EINECS: 219-679-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...