2-Ethylhexyl diphenyl phosphate
Leverandør: EHRENSTORFER GMBH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
EHERC13342300EA 1260 DKK
EHERC13342300
2-Ethylhexyl diphenyl phosphate
2-Ethylhexyl diphenyl phosphate
Formel: C₂₀H₂₇O₄P
MDL Number: MFCD00059949
CAS-nummer: 1241-94-7
EINECS: 214-987-2
UN nummer: 3082
ADR: 9,III