Barium chlorate monohydrate ≥98%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

A10128.0EEA 1790 DKK
A10128.0E
Barium chlorate monohydrate ≥98%
Barium chlorate monohydrate
Formel: BaCl₂O₆·1H₂O
Molekylvægt: 322,26 g/mol (304,24 anhy)
Smeltepunkt: 414 °C
Massefylde: 3,18
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149147
CAS-nummer: 10294-38-9
EINECS: 236-760-7
UN nummer: 1445
ADR: 5.1,II

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR