4-Phenoxybenzophenone ≥95.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAP1328-5GEA
879
DKK
TCIAP1328-5G
4-Phenoxybenzophenone ≥95.0% (by GC)
4-Phenoxybenzophenone
Formel:
C₁₉H₁₄O₂ |
MDL Number:
MFCD00017553 CAS-nummer: 6317-73-3 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...