Benzophenone-4,4'-dicarboxylic acid ≥95.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAB1741-5GEA
936
DKK
TCIAB1741-5G
Benzophenone-4,4'-dicarboxylic acid ≥95.0% (by HPLC, titration analysis)
Benzophenone-4,4'-dicarboxylic acid
Formel:
C₁₅H₁₀O₅ |
MDL Number:
MFCD00016538 CAS-nummer: 964-68-1 |
Specifikationer Testresultater
Appearance | White to Light yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...