Diphenylacetaldehyde ≥95.0% (by GC)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAD2492-25GEA
3220
DKK
TCIAD2492-25G
TCIAD2492-5G
Diphenylacetaldehyde ≥95.0% (by GC)
Diphenylacetaldehyde
Formel:
C₁₄H₁₂O Molekylvægt: 196,2400 g/mol Kogepunkt: 175°C |
MDL Number:
MFCD00006972 CAS-nummer: 947-91-1 EINECS: 213-433-7 |
![]() |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Red clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...