N-(4-Methoxybenzylidene)aniline ≥98%

Leverandør: Thermo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

L01951.14EA 1300 DKK
L01951.14
N-(4-Methoxybenzylidene)aniline ≥98%
N-(4-Methoxybenzylidene)aniline
Formel: C₁₄H₁₃NO
Molekylvægt: 211,26 g/mol
Smeltepunkt: 61…64 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00025836
CAS-nummer: 836-41-9
EINECS: 212-647-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR