N-Cyclopropyl-4-boronobenzenesulphonamide ≥98%
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H53170.MDEA
1170
DKK
H53170.MD
H53170.03
N-Cyclopropyl-4-boronobenzenesulphonamide ≥98%
N-Cyclopropyl-4-boronobenzenesulphonamide
Formel:
C₉H₁₂BNO₄S Molekylvægt: 241,07 g/mol Smeltepunkt: 142…144 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD07363755 CAS-nummer: 871329-67-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...