3-Boronobenzenesulphonamide ≥97%
Leverandør: Thermo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
H52628.MDEA
5610
DKK
H52628.MD
3-Boronobenzenesulphonamide ≥97%
3-Boronobenzenesulphonamide
Formel:
C₆H₈BNO₄S Molekylvægt: 201,00 g/mol Smeltepunkt: 260…266 °C |
MDL Number:
MFCD06659840 CAS-nummer: 850568-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...