(+)-Taxifolin

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

ROTHT893.1EA 3273.14 DKK
ROTHT893.1
(+)-Taxifolin
(+)-Taxifolin
Formel: C₁₅H₁₂O₇
Molekylvægt: 304,2516 g/mol
MDL Number: MFCD00006845
CAS-nummer: 480-18-2
EINECS: 207-543-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR