Sodium acetate-[1-¹³C] (99% ¹³C )

Leverandør: CAMBRIDGE ISOTOPE LABS., INC.
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

CAMBCLM-156-10EA 0 DKK
CAMBCLM-156-10 CAMBCLM-156-5
Sodium acetate-[1-¹³C] (99% ¹³C )
Sodium acetate-[1-¹³C]
Formel: H₃CCOONa
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00037666
CAS-nummer: 23424-28-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR