Guanine hydrochloride ≥98.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAG0170-25GEA
476
DKK
TCIAG0170-25G
Guanine hydrochloride ≥98.0% (by titrimetric analysis)
Guanine hydrochloride
Formel:
C₅H₆ClN₅O Molekylvægt: 187,59 g/mol |
MDL Number:
MFCD00213670 CAS-nummer: 635-39-2 EINECS: 211-235-5 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % (calcd.on dried substance) |
Drying loss | max. 10.0 % |
Solubility in hot dilute HCl | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...