Anisomycin (from Streptomyces griseolus), Millipore®
Leverandør: Merck Millipore Calbiochem
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
176880-10EA 795 DKK
176880-10
Anisomycin (from Streptomyces griseolus), Millipore®
Anisomycin
Formel: C₁₄H₁₉NO₄
MDL Number: MFCD00077650
CAS-nummer: 22862-76-6
EINECS: 245-269-7
UN nummer: 3462
ADR: 6.1,III