10,12-Heptadecadiynoic acid ≥97.0% (by titrimetric analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAH0775-100MGEA
2130
DKK
TCIAH0775-100MG
10,12-Heptadecadiynoic acid ≥97.0% (by titrimetric analysis)
10,12-Heptadecadiynoic acid
Formel:
C₁₇H₂₆O₂ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00142348 CAS-nummer: 28393-06-8 |
![]() |
Specifikationer Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 40.0 to 44.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...