8-Chlorotheophylline ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
TCIAC0293-250GEA
928
DKK
TCIAC0293-250G
TCIAC0293-25G
8-Chlorotheophylline ≥98.0% (by HPLC, titration analysis)
8-Chlorotheophylline
Formel:
C₇H₇ClN₄O₂ |
MDL Number:
MFCD00005581 CAS-nummer: 85-18-7 EINECS: 201-590-4 |
![]() |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in Sodium hydroxide solution | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...