8-Bromotheophylline ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TCIAB3456-5GEA
2290
DKK
TCIAB3456-5G
TCIAB3456-1G
8-Bromotheophylline ≥98.0% (by HPLC, titration analysis)
8-Bromotheophylline
Formel:
C₇H₇BrN₄O₂ |
MDL Number:
MFCD00022664 CAS-nummer: 10381-75-6 EINECS: 233-846-6 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...