Hydrochlorothiazide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

PROOMM0011.00EA 0 DKK
PROOMM0011.00
Hydrochlorothiazide
Hydrochlorothiazide
Formel: C₇H₈ClN₃O₄S₂
MDL Number: MFCD00051765
CAS-nummer: 58-93-5
EINECS: 200-403-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR