Retinoic acid

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TICIUSP1674004
Retinoic acid
Retinoic acid
Formel: C₂₀H₂₈O₂
Molekylvægt: 300,44 g/mol
MDL Number: MFCD00001551
CAS-nummer: 302-79-4
EINECS: 206-129-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR