Hesperidin
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ROTH5800.1EA
502.99
DKK
ROTH5800.1
Hesperidin
Hesperidin
Formel:
C₂₈H₃₄O₁₅ Molekylvægt: 610,56 g/mol |
MDL Number:
MFCD00075663 CAS-nummer: 520-26-3 EINECS: 208-288-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...