Tin(IV) phthalocyanine dichloride
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAP0997-1GEA
870
DKK
TCIAP0997-1G
Tin(IV) phthalocyanine dichloride
Tin(IV) phthalocyanine dichloride
Formel:
C₃₂H₁₆Cl₂N₈Sn Molekylvægt: 702,139 g/mol |
MDL Number:
MFCD00054210 CAS-nummer: 18253-54-8 UN nummer: 3146 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | Dark red to Dark purple to Dark blue powder to crystal |
Ignition residue (as SnO2) | 20.0 to 25.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...