Diclofenac sodium salt, Millipore®
Leverandør: Merck Millipore Calbiochem
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
287840-1EA
658
DKK
287840-1
Diclofenac sodium salt, Millipore®
Diclofenac sodium salt
Formel:
C₁₄H₁₀Cl₂NNaO₂ Molekylvægt: 318,1 g/mol |
MDL Number:
MFCD00082251 CAS-nummer: 15307-79-6 EINECS: 239-346-4 UN nummer: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...