Diclofenac ≥98.0% (by GC, titration analysis)
Leverandør: TCI EUROPE
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
TCIAD3748-25GEA
567
DKK
TCIAD3748-25G
Diclofenac ≥98.0% (by GC, titration analysis)
Diclofenac
Formel:
C₁₄H₁₁Cl₂NO₂ Smeltepunkt: 158°C |
CAS-nummer:
15307-86-5 EINECS: 239-348-5 UN nummer: 2811 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...