N-Phenylurethane ≥99.0% (by GC)

Leverandør: TCI EUROPE
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

TCIAP1090-25GEA 238 DKK
TCIAP1090-25G TCIAP1090-500G
N-Phenylurethane ≥99.0% (by GC)
N-Phenylurethane
Formel: C₉H₁₁NO₂
Molekylvægt: 165,19
Kogepunkt: 237°C
MDL Number: MFCD00026806
CAS-nummer: 101-99-5
EINECS: 202-995-9
https://dk.vwr.com/stibo/web/std.lang.all/17/21/28991721.jpg

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 49.0 to 52.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR