Glass disposal boxes, Azlon®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Azlon®
AZLODBG001PEA 904 DKK
AZLODBG001P AZLODBG002P
Glass disposal boxes, Azlon®
Bokse Affaldsbeholdere
Cardboard.
  • Self assembly rigid boxes for safe disposal of broken glass
  • Glass is safely held in strong leak-proof and puncture resistant LDPE liner
  • Lid has an integral waste flap for easy use and a sealing feature for use when full
These boxes are clearly printed with instructions for use.