Dishes, evaporating
Leverandør: Semadeni
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SEMA0314EA 227 DKK
SEMA0314
Dishes, evaporating
Bakker/Tallerkener/Potter Fordampningsbassiner/skåle
Made of PP, translucent, with spout.
  • Very good temperature resistance
  • Very good chemical resistance
  • Easy to clean