Wide opening screw neck vials, 9 mm, SureStop™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SureStop™
NSCAC5000-592EA 403.5 DKK
NSCAC5000-2 NSCAC5000-592 NSCAC5000-592W NSCAC5000-2W NSCAC5000-1
Wide opening screw neck vials, 9 mm, SureStop™
Hætteglas
As an integral component of AVCS technology, SureStop™ vials offer the sealing and performance characteristics of a crimp top vial and the versatility properties of a threaded vial.
  • Optimal septum compression across the opening of the vial
  • Design allows the positioning of the closure to be consistently "flat" producing a more cylindrical vial profile, which improves autosampler recognition and compatibility
  • Designed for use with nearly every autosampler on the market
This is achieved by incorporating a definite stop point into the design of the vial finish, preventing over tightening of the closure. SureStop™ vials remove all doubts about the degree of tightening necessary for optimal sealing.