Stereo zoom microscopes, SMZ-161
Leverandør: Motic
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
630-2029EA 5150 DKK
MOTI1100201300022 MOTI1100201300032 MOTI1100201300062 MOTI1100201300082
Stereo zoom microscopes, SMZ-161
The SMZ-161 offer improved optical performance.
  • Reflector of transmitted light providing clear, distortion-free, 3-D images
  • L models feature integrated halogen light
  • C-mount adapter on trinocular models
Designed for industrial applications, especially to observe assembly and inspect small parts or integrated circuits on PCB, LCD production line.
Bestillingsoplysninger: 0,5×, 0,65×, 1× adapters are available.