Nitrogen generators, Infinity NM range
Leverandør: Peak Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
564-0117EA 0 DKK
564-0117 564-0116
Nitrogen generators, Infinity NM range
Gasgeneratorer Nitrogengeneratorer
These generators are designed to be connected to an existing air supply and can cater for the requirements of LC-MS, LC-MS-MS, sample evaporators, ELSD and NMR applications.
Inlet air requirement: Infinity NM18L 50 l/min; Infinity NM32L 90 l/min
Min./max. air inlet pressure: 125 to 145 psi
Min./max. operating temperature: 5 to 30 °C
Pressure dew point: −40 °C