Polybuffer for chromatofocusing
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17-0713-01EA 2359 DKK
GEHE17-0713-01 GEHE17-0714-01
Polybuffer for chromatofocusing
Polybuffers are used for chromatofocusing and the buffer forms a linear pH gradient after titration on Mono P columns. Chromatofocusing is a powerful protein separation technique separating proteins by isoelectric point (pI).
  • Resolves pI differences of 0,04 pH units
  • Mixtures of selected amphoteric buffering substances of different pI and pKa values
  • Polybuffer 74 for any pH gradient between 7 and 4
  • Polybuffer 96 for pH gradients that should begin above pH 7