Precast gels, CleanGel IEF
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CleanGel™
18-1035-32EA 4060 DKK
18-1035-32
Precast gels, CleanGel IEF
Elektroforese geler Forstøbte Geler
CleanGel IEF is a dried polyacrylamide gel optimised for analytical isoelectric focusing (IEF). The gel is excellent for separating enzymes with preserved activity.
The gel is cast on a polyester support film and carefully washed to remove catalysts and nonpolymerized substances and then dried. Prior to isoelectric focusing, the dried gel is rehydrated in a solution containing Pharmalyte™ carrier ampholytes. Additives such as urea and/or detergents can also be added at this stage. CleanGel IEF is rehydrated in a flat tray (GelPool) to a thickness of 0,43 mm.
  • Perform sensitive procedures using precast gels that are carefully washed and dried after casting to remove catalysts and unpolymerised material
  • Customise IEF separations by reswelling in a suitable mixture of carrier ampholytes and other additives such as detergents or denaturants
  • Handle mechanically stable, plastic-backed gels with confidence
The gel is cast on a polyester support film and carefully washed to remove catalysts and nonpolymerized substances and then dried. Prior to isoelectric focusing, the dried gel is rehydrated in a solution containing Pharmalyte™ carrier ampholytes. Additives such as urea and/or detergents can also be added at this stage. CleanGel IEF is rehydrated in a flat tray (GelPool) to a thickness of 0,43 mm.
Introducing carrier ampholytes and additives into the gel after the polymerisation eliminates the risk of copolymerising chemicals in the gel matrix. Thus, CleanGel IEF gels give low electroendosmosis, resulting in sharp bands and minimum pH gradient drift. Even additives that are sensitive to polymerisation reactions, or that inhibit the polymerisation can be used.