GEHE17-1332-01
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17-1332-01EA 911.88 DKK
17-1332-01
GEHE17-1332-01
PlusOne™ Repel-Silane ES is a 2% solution of dimethyldichlorosilane dissolved in octamethylcyclooctasilane, a low toxicity, environmentally safe solvent. It is used to inhibit sticking of polyacrylamide gels, agarose gels and nucleic acids to glass surfaces.