PlusOne™ Acrylamide IEF, ReadySol
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17-1310-01EA 2127 DKK
17-1310-01
PlusOne™ Acrylamide IEF, ReadySol
Elektroforese geler Agarose og Akrylamid
PlusOne™ Acrylamide IEF is outstanding for use in isoelectric focusing and DNA sequencing. In addition to the absence of impurities that interfere with polymerisation, acrylamide IEF is essentially free of charged acrylamide derivatives (<20 ppm) for very stable pH gradients. ReadySol, a solution of acrylamide and bisacrylamide, is convenient and fast with minimal preparation and handling. ReadySol ensures a reproducible solution composition.