Desiccator plates
Leverandør: Vitlab
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VITL65975EA 1700 DKK
VITL80551 VITL80553 VITL65975 VITL65965
Desiccator plates
Porcelain plates are suitable for hot objects. PP plates are usable up to a maximum of 120 °C.