VWR® TBS/Tween® 20X Washing Buffer, Liquid Concentrate, UltraPure Grade
Leverandør: Avantor
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
K873-1L
VWR® TBS/Tween® 20X Washing Buffer, Liquid Concentrate, UltraPure Grade
Buffere Elektroforese buffere Washing Buffers
Convenient liquid concentrate that produces 10 litres of 1X buffer for blotting and ELISA applications.
  • Prevents high background by effectively removing unbound reagents