Leak diverter for roofs, PIG®
Leverandør: New Pig
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PIG®
NEWPTLS564-YWEA 0 DKK
NEWPTLS564-YW
Leak diverter for roofs, PIG®
Spild materialer
Leak diverter with telescopic, aluminium pole which extends into place to install the leak diverter up to 6 metres in the air without a ladder.
  • Tension mounted pole with rubber cups secures pole to floor and ceiling
  • Makes it easy for one person to install or move the diverter
  • Non permeable tarpaulin is vinyl coated for strength and durability
  • Resists punctures, UV and mildew for long life
Bestillingsoplysninger: Hose sold separately.