Nuclease mix
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
80-6501-42EA 921 DKK
80-6501-42
Nuclease mix
Protein oprensningssystemer
Nuclease mix offers an effective mix of DNase and RNase enzymes, as well as the necessary cofactors for optimal nuclease activity.
  • Specifically developed for removal of nucleic acids in sample preparation for IEF/2-D electrophoresis applications in proteomic studies
  • Compatible with Cytiva protease inhibitor mix
  • Ready to use: Simply add the mix to the sample and incubate
  • Does not contain EDTA, which is an inhibitor of nuclease
  • Colorless
  • DIGE
Removal of nucleic acids is often required to avoid contamination and subsequent artifacts on 2-D gels. Specifically developed for removal of nucleic acids in sample preparation for IEF/2-D electrophoresis applications in proteomic studies.
The protease inhibitor mix contains an optimized concentration of competitive and noncompetitive protease inhibitors, which protect proteins during purification from animal and plant tissues, as well as yeast and bacteria. It effectively inhibits over 95% of protease activity.
Advarsel: Store at −20°C