Ultra-accurate digital thermometers
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-1929EA 547 DKK
VWRI620-1929
Ultra-accurate digital thermometers
These ready to use thermometers are designed for highly accurate, quality temperature management. They can be used in air, gas, liquids and solids. They are suitable for pharmaceutical and biotechnology companies, food industry, government institutes and environment.
  • Low battery warning indicator
  • Calibratable: Offset and slope input
  • 'HOLD' function
  • All probes are permanently connected to device
  • Housing is made of impact resistant ABS
  • Battery life: approx. 200 operating hours
Leveringsinformation: Supplied with 9 volt battery, probe and manual.