Affinity chromatography columns, TriCorn™ Mono P™
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Tricorn™
17-5171-01EA 17309 DKK
GEHE17-5171-01
Affinity chromatography columns, TriCorn™ Mono P™
Mono P™ are Tricorn™ high performance columns. The columns are pre-packed glass columns for high performance chromatofocusing of proteins, peptides, polynucleotides and other biomolecules. Mono P™ 5/200 GL offers high selectivity for biomolecules.
  • Greatly concentrates samples and resolves pI differences of 0,02 pH units
  • Monodisperse 10 µm porous beads yield high resolution, dynamic capacity, reproducibility, and durability