Anion exchange chromatography column, Mono Q HR
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17-0506-01EA 131610 DKK
17-0506-01
Anion exchange chromatography column, Mono Q HR
Kromatografikolonner
Mono Q HR is a strong anion exchanger prepacked with MonoBeads™. The column is an excellent choice for the polishing step in purification of proteins, peptides, and other biomolecules when high purity is required.
  • Monodisperse 10 µm porous beads yield high resolution, dynamic capacity, reproducibility, and durability
  • Maximum resolution is achieved from the efficiency of small, perfectly spherical monodispersed particles optimally packed in columns coupled with the excellent selectivities of the Q ion exchangers
  • Compatible with ÄKTA design, FPLC System, and other high-performance LC systems
Not suitable for use with ÄKTAprime plus system.