Ion exchange chromatography columns, Mini Q PC
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17-0686-01EA 17233 DKK
GEHE17-0686-01
Ion exchange chromatography columns, Mini Q PC
Mini Q PC precision columns are prepacked with MiniBeads™ for microscale final polishing and analytical ion exchange separations.
  • Monodisperse 3 µm nonporous beads yield exceptional resolution, speed, reproducibility and durability
  • PC columns are designed for micropurification
  • Nonporous matrix of MiniBeads™ improves resolution and flow kinetics, better withstands back pressure, and minimises nonspecific adsorption of the sample
  • Not suitable for use with ÄKTAprime plus system
Bestillingsoplysninger: A precision column holder needs to be ordered separately when used with standard chromatography systems.