Precast gels, PhastGel® IEF
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17-0543-01EA 1314 DKK
17-0543-01
Precast gels, PhastGel® IEF
Elektroforese geler Forstøbte Geler
PhastGel® IEF media are precast homogeneous (5% T, 3% C) polyacrylamide gels containing 2% to 6% pharmalyte as carrier ampholyte. Pharmalyte generates stable, linear pH gradients with uniform conductivity across the entire pH range, allowing field strengths >500 V/cm for high resolution separations.
  • Separate proteins and peptides on denaturing, native, and isoelectric focusing gels
  • Save time and improve reproducibility with precast gels, buffer strips and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready to use silver staining kits