VWR® LaPrep Sigma, Preparative HPLC System
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR® LaPrep
905-0550EA 0 DKK
905-0550 905-0509 905-0508 905-0507 905-0528 905-0505 905-0527 905-0504 905-0515 905-0526 905-0514 905-0503 905-0525 905-0502 905-0524 905-0523 905-0512 905-0501 905-0500 905-0511 905-0510
VWR® LaPrep Sigma, Preparative HPLC System
Kromatografisystemer Komplette Preparative HPLC Systemer
The LaPrep Sigma is a highly robust and compact preparative chromatography system running on the intuitive EZChrom platform. Being a truly modular system, with many possible configurations, it has one of the smallest footprints in the market (242×352 mm). With an easy to use touch screen panel and Ethernet connection, it is an ideal addition to any laboratory needing a flexible preparative HPLC solution.
The LaPrep Sigma has capabilities ranging from analytical to preparative scale, pump capacity from 1 to 600 ml/min that can be easily connected to a fully automated fraction collector. Along with the options of a single- or multiple-wavelength UV detector, and autosampler or single-injection sample delivery, the LaPrep Sigma is an excellent choice for any preparative laboratory whatever the configuration requirements.
Key to the flexibility of LaPrep Sigma is the assistant module. The assistant houses the fractionation and injection valve plus automated injection pump, meaning that minimal bench space is used. A variety of configuration options are available.
For further information, demo requests, or application support, please get in touch with your local Avantor chromatography specialist or email chromatography@eu.vwr.com.